missing translation for 'onlineSavingsMsg'
Learn More

Methyl (2S)-2-(2-Chlorophenyl)-2-[2-(thiophen-2-yl)ethylamino]acetate Hydrochloride, Mikromol™

Product Code. 30050688
Click to view available options
Quantity:
25 mg
Unit Size:
25mg
This item is not returnable. View return policy

Product Code. 30050688

Brand: Mikromol MM0455.220025

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Discover Mikromol - your trusted source for high-quality pharmaceutical reference standards. Supporting accurate, compliant analysis with ISO 17034-accredited materials designed for confidence in every result.

Methyl (2S)-2-(2-Chlorophenyl)-2-[2-(thiophen-2-yl)ethylamino]acetate Hydrochloride
TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material (S)-(+)-N-(2-(2-Thienyl)ethyl)-2-chlorophenyl Glycine methyl ester hydrochloride
CAS 141109-19-5
Analyte or Component Names (S)-(+)-N-(2-(2-Thienyl)ethyl)-2-chlorophenyl Glycine methyl ester hydrochloride
Shipping Condition Room Temperature
Molecular Formula C15 H16 Cl N O2 S . Cl H
Certifications/Compliance ISO 9001
Synonym Benzeneacetic acid, 2-chloro-α-[[2-(2-thienyl)ethyl]amino]-, methyl ester, hydrochloride (1:1), (αS)-, Benzeneacetic acid, 2-chloro-α-[[2-(2-thienyl)ethyl]amino]-, methyl ester, hydrochloride, (S)-, Benzeneacetic acid, 2-chloro-α-[[2-(2-thienyl)ethyl]amino]-, methyl ester, hydrochloride, (αS)- (9CI), (+)-(S)-Methyl α-[[2-(2-thienyl)ethyl]amino]-α-(2-chlorophenyl)acetate hydrochloride, (+)-Methyl α-(2-thienylethamino)-α-(2-chlorophenyl)acetate hydrochloride, (S)-(+)-Methyl α-[[2-(thien-2-yl)ethyl]amino]-α-(2-chlorophenyl)acetate hydrochloride
InChI Formula InChI=1S/C15H16ClNO2S.ClH/c1-19-15(18)14(12-6-2-3-7-13(12)16)17-9-8-11-5-4-10-20-11;/h2-7,10,14,17H,8-9H2,1H3;1H/t14-;/m0./s1
SMILES Cl.COC(=O)[C@@H](NCCc1cccs1)c2ccccc2Cl
IUPAC Name methyl (2S)-2-(2-chlorophenyl)-2-(2-thiophen-2-ylethylamino)acetate;hydrochloride
Molecular Weight (g/mol) 346.27
Quantity 25 mg
Formula Weight 345.0357 g/mol
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.