missing translation for 'onlineSavingsMsg'
Learn More

Methyl (2RS)-1,4-Benzodioxane-2-carboxylate, Mikromol™

Product Code. 17911986
Change view
Click to view available options
Quantity:
25 mg
Unit Size:
25mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
17911986 25 mg 25mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 17911986 Supplier Mikromol Supplier No. MM0386.110025
 Restricted Item
Estimated Shipment: 13-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

Discover Mikromol - your trusted source for high-quality pharmaceutical reference standards. Supporting accurate, compliant analysis with ISO 17034-accredited materials designed for confidence in every result.

Methyl (2RS)-1,4-Benzodioxane-2-carboxylate
TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Methyl (2RS)-1,4-Benzodioxane-2-carboxylate
CAS 3663-79-4
Analyte or Component Names Methyl (2RS)-1,4-Benzodioxane-2-carboxylate
Shipping Condition Room Temperature
Molecular Formula C10 H10 O4
Certifications/Compliance ISO 9001
Synonym 1,4-Benzodioxin-2-carboxylic acid, 2,3-dihydro-, methyl ester, 1,4-Benzodioxan-2-carboxylic acid, methyl ester (6CI,7CI,8CI), Methyl 1,4-benzodioxan-2-carboxylate, Methyl 1,4-benzodioxane-2-carboxylate, Methyl (2RS)-1,4-benzodioxane-2-carboxylate
InChI Formula InChI=1S/C10H10O4/c1-12-10(11)9-6-13-7-4-2-3-5-8(7)14-9/h2-5,9H,6H2,1H3
SMILES COC(=O)C1COc2ccccc2O1
IUPAC Name methyl 2,3-dihydro-1,4-benzodioxine-3-carboxylate
Molecular Weight (g/mol) 194.18
Quantity 25 mg
Formula Weight 194.0579 g/mol
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.