missing translation for 'onlineSavingsMsg'
Learn More

(4R,5S)-(+)-4-Methyl-5-phenyl-2-oxazolidinone, 99%

Product Code. 10061981 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
Packaging:
Glass Bottle
Unit Size:
1g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10061981 1 g Glass Bottle 1g
10698444 5 g Glass Bottle 5g
10434765 25 g Glass Bottle 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 10061981 Supplier Thermo Scientific Acros Supplier No. 337630010
 Restricted Item
Estimated Shipment: 29-07-2026
Add to basket
This item is not returnable. View return policy

CAS: 77943-39-6 | C10H11NO2 | 177.2 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 77943-39-6
Molecular Formula C10H11NO2
Molecular Weight (g/mol) 177.2
MDL Number MFCD00010845
InChI Key PPIBJOQGAJBQDF-VXNVDRBHSA-N
Synonym 4r,5s-+-4-methyl-5-phenyl-2-oxazolidinone, 4r,5s-4-methyl-5-phenyloxazolidin-2-one, 4r,5s-4-methyl-5-phenyl-1,3-oxazolidin-2-one, 2-oxazolidinone, 4-methyl-5-phenyl-, 4r,5s, 4r,5s-4-methyl-5-phenyl-2-oxazolidinone, meph-2-oxazolidinone, pubchem6871, 2-oxazolidinone, 4-methyl-5-phenyl-, 4r-cis, 4-methyl-5-phenyl-2-oxazolidinone 4r-cis, 4alpha-methyl-5alpha-phenyl-2-oxazolidinone
PubChem CID 2733812
IUPAC Name (4R,5S)-4-methyl-5-phenyl-1,3-oxazolidin-2-one
SMILES CC1C(OC(=O)N1)C2=CC=CC=C2

Specifications

Melting Point 118°C to 121°C
CAS Min % 98.5
CAS Max % 100.0
Color Brown-Yellow to Colorless
Infrared Spectrum Authentic
Assay Percent Range 98.5% min. (HPLC)
Packaging Glass Bottle
Specific Rotation + 162
Specific Rotation Condition + 162.00 (20.00°C c=2,CHCL3)
Quantity 1 g
Formula Weight 177.2
Percent Purity 99%
Physical Form Liquid
Chemical Name or Material (4R, 5S)-(+)-4-Methyl-5-phenyl-2-oxazolidinone
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.