missing translation for 'onlineSavingsMsg'
Learn More

Methyl (2R,3S)-3-benzamido-2-hydroxy-3-phenylpropionate, 98%

Product Code. p-7053892
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
This item is not returnable. View return policy

Product Code. 15446666

Brand: Thermo Scientific Alfa Aesar H61641.03

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 32981-85-4 | C17H17NO4 | 299.326 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 32981-85-4
Molecular Formula C17H17NO4
Molecular Weight (g/mol) 299.326
MDL Number MFCD00673331
InChI Key UYJLJICUXJPKTB-LSDHHAIUSA-N
Synonym 2r,3s-methyl 3-benzamido-2-hydroxy-3-phenylpropanoate, methyl 2r,3s-3-benzoylamino-2-hydroxy-3-phenylpropanoate, bz-rs-iser 3-ph-ome, benzenepropanoic acid, beta-benzoylamino-alpha-hydroxy-, methyl ester, alphar,betas, methyl 2r,3s-3-benzamido-2-hydroxy-3-phenylpropanoate, 2r,3s-n-tert-butoxycarbony-3-phenylisoserine methyl ester, methyl 2r,3s-2-hydroxy-3-phenyl-3-phenylformamido propanoate, pubchem9699, n-benzoyl-2r,3s-3-phenylisoserine methyl ester, methyl 2r,3s-n-benzoylphenylisoserinate
PubChem CID 182104
IUPAC Name methyl (2R,3S)-3-benzamido-2-hydroxy-3-phenylpropanoate
SMILES COC(=O)C(C(C1=CC=CC=C1)NC(=O)C2=CC=CC=C2)O

Specifications

Melting Point 178°C to 183°C
Color White
Quantity 1 g
Formula Weight 299.32
Percent Purity 98%
Physical Form Crystalline Powder
Chemical Name or Material Methyl (2R,3S)-3-benzamido-2-hydroxy-3-phenylpropionate

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.