missing translation for 'onlineSavingsMsg'
Learn More

Methyl 2-(trifluoromethyl)pyrimidine-4-carboxylate, 97%, Thermo Scientific Chemicals

Product Code. p-7035110
Change view
Click to view available options
Quantity:
250 mg
1 g
Unit Size:
1g
250mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15460136 250 mg 250mg
15450136 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15460136 Supplier Thermo Scientific Alfa Aesar Supplier No. H52818.MD

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 878745-51-8 | Molecular Formula: C7H5F3N2O2 | Molecular Weight: 206.12 g/mol

Methyl 2-(trifluoromethyl)pyrimidine-4-carboxylate is used as pharmaceutical intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Methyl 2-(trifluoromethyl)pyrimidine-4-carboxylate is used as pharmaceutical intermediate.

Notes
Stable under recommended storage conditions. Store away from oxidizing agents.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 878745-51-8
Molecular Formula C7H5F3N2O2
Molecular Weight (g/mol) 206.12
MDL Number MFCD05662690
InChI Key VBFLWQBZJODKRL-UHFFFAOYSA-N
Synonym methyl 2-trifluoromethyl pyrimidine-4-carboxylate, 2-trifluoromethyl-pyrimidine-4-carboxylic acid methyl ester, methyl2-trifluoromethyl pyrimidine-4-carboxylate, methyl 2-trifluoromethyl-4-pyrimidine carboxylate, 4-pyrimidinecarboxylic acid, 2-trifluoromethyl-, methyl ester, 4-pyrimidinecarboxylicacid, 2-trifluoromethyl-, methyl ester, 4-pyrimidinecarboxylicacid,2-trifluoromethyl-,methyl ester
PubChem CID 17750127
IUPAC Name methyl 2-(trifluoromethyl)pyrimidine-4-carboxylate
SMILES COC(=O)C1=NC(=NC=C1)C(F)(F)F

Specifications

Quantity 250 mg
Formula Weight 206.12
Percent Purity 97%
Chemical Name or Material Methyl 2-(trifluoromethyl)pyrimidine-4-carboxylate

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.