missing translation for 'onlineSavingsMsg'
Learn More

Methyl 2-piperidinoisonicotinate, 97%, Thermo Scientific™

Product Code. 10142794
Change view
Click to view available options
Quantity:
250 mg
1 g
Packaging:
Amber Glass Bottle
Unit Size:
1g
250mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10142794 1 g Amber Glass Bottle 1g
10173124 250 mg Amber Glass Bottle 250mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10142794 Supplier Thermo Scientific Maybridge Supplier No. CC50322DA
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Chemical Identifiers

CAS 888070-05-1
Molecular Formula C12H16N2O2
Molecular Weight (g/mol) 220.27
MDL Number MFCD08741429
InChI Key CCOBIXWZODVNKL-UHFFFAOYSA-N
Synonym methyl 2-piperidinoisonicotinate, methyl 2-piperidin-1-ylpyridin-4-ylcarboxylate, methyl 2-piperidin-1-yl pyridine-4-carboxylate, methyl 2-piperidin-1-yl isonicotinate, methyl 2-piperidin-1-ylisonicotinate, methyl 2-piperidylpyridine-4-carboxylate, 4-pyridinecarboxylicacid, 2-1-piperidinyl-, methyl ester, 4-pyridinecarboxylicacid,2-1-piperidinyl-,methyl ester
PubChem CID 18525843
IUPAC Name methyl 2-piperidin-1-ylpyridine-4-carboxylate
SMILES COC(=O)C1=CC(=NC=C1)N1CCCCC1

Specifications

CAS Min % 96.5
CAS Max % 100.0
Assay Percent Range 97%
Packaging Amber Glass Bottle
Quantity 1 g
Formula Weight 220.27
Percent Purity 97%
Chemical Name or Material Methyl 2-piperidinoisonicotinate
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.