missing translation for 'onlineSavingsMsg'
Learn More

Methyl 2-oxocyclohexanecarboxylate, 90%

Product Code. 10651555 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
10 mL
50 mL
Packaging:
Glass bottle
Unit Size:
10mL
50mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10651555 50 mL Glass bottle 50mL
10129132 10 mL Glass bottle 10mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10651555 Supplier Thermo Scientific Acros Supplier No. 367010500
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 41302-34-5 | C8H12O3 | 156.2 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 41302-34-5
Molecular Formula C8H12O3
Molecular Weight (g/mol) 156.2
MDL Number MFCD00077699
InChI Key JEENWEAPRWGXSG-UHFFFAOYSA-N
Synonym methyl 2-oxocyclohexanecarboxylate, 2-methoxycarbonylcyclohexanone, 2-carbomethoxycyclohexanone, cyclohexanecarboxylic acid, 2-oxo-, methyl ester, methyl cyclohexanone-2-carboxylate, 2-oxo-cyclohexanecarboxylic acid methyl ester, 2-hydroxy methyl cyclohexanone, 2-oxocyclohexanecarboxylic acid, methyl ester, pubchem20380, acmc-1ao3u
PubChem CID 170489
IUPAC Name methyl 2-oxocyclohexane-1-carboxylate
SMILES COC(=O)C1CCCCC1=O

Specifications

Color Colorless to Yellow
Density 1.1090g/mL
Boiling Point 50°C (0.2 mmHg)
Flash Point 85°C
Infrared Spectrum Authentic
Assay Percent Range 88% min. may occur as tautomers (GC)
Packaging Glass bottle
Refractive Index 1.4830 to 1.486
Quantity 50 mL
Beilstein 10, IV, 2606
Specific Gravity 1.109
Formula Weight 156.2
Percent Purity 90%
Physical Form Liquid
Chemical Name or Material Methyl 2-oxocyclohexanecarboxylate
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.