missing translation for 'onlineSavingsMsg'
Learn More

Methyl 2-Octynoate, TRC

Product Code. 30470519
Click to view available options
Quantity:
1 g
Unit Size:
1g
This item is not returnable. View return policy

Product Code. 30470519

Brand: Toronto Research Chemicals TRCM3244951G

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Methyl-2-Octynoate
CAS 111-12-6
Molecular Formula C9 H14 O2
Quantity 1 g
Synonym 2-Octynoic acid-methyl ester, 2-Octynoic acid','methyl ester (6CI','7CI','8CI','9CI','ACI), Folione, Methyl 2-octynate, Methyl 2-octynoate, Methyl pentylacetylenecarboxylate, NSC 72098, Vert de violette','artificial
InChI Formula InChI=1S/C9H14O2/c1-3-4-5-6-7-8-9(10)11-2/h3-6H2,1-2H3
SMILES CCCCCC#CC(=O)OC
IUPAC Name methyl oct-2-ynoate
Molecular Weight (g/mol) 154.21
Formula Weight 154.0994
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.