missing translation for 'onlineSavingsMsg'
Learn More

Methyl 2-nitrobenzoate, 99%, Thermo Scientific™

Product Code. 10710871 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
25 g
Packaging:
Glass bottle
Unit Size:
25g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10710871 25 g Glass bottle 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 10710871 Supplier Thermo Scientific Acros Supplier No. 168430250
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Specifications

CAS 606-27-9
Color Brown-Orange to Yellow
Density 1.2800g/mL
Boiling Point 95.0°C (1.0 mmHg)
Flash Point >112°C
Infrared Spectrum Authentic
Assay Percent Range 99%
Packaging Glass bottle
Molecular Formula C8H7NO4
Linear Formula O2NC6H4CO2CH3
Refractive Index 1.5340 to 1.5360
MDL Number MFCD00007136
Quantity 25 g
Beilstein 09, 372
Specific Gravity 1.28
Synonym methyl o-nitrobenzoate, benzoic acid, 2-nitro-, methyl ester, 2-nitro-benzoic acid methyl ester, benzoic acid, o-nitro-, methyl ester, 2-nitrobenzoic acid methyl ester, methyl-2-nitrobenzoate, methyl o-nitro benzoate, methyl ortho-nitrobenzoate, acmc-209ml4, nitrobenzoic acid methyl ester
InChI Key AOXPHVNMBPFOFS-UHFFFAOYSA-N
SMILES COC(=O)C1=CC=CC=C1[N+](=O)[O-]
IUPAC Name methyl 2-nitrobenzoate
Molecular Weight (g/mol) 181.147
PubChem CID 69072
Formula Weight 181.15
Percent Purity 99%
Physical Form Liquid
Chemical Name or Material Methyl 2-nitrobenzoate, 99%
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.