missing translation for 'onlineSavingsMsg'
Learn More

Methyl 2-naphthoate, 99%

Product Code. 11382377
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11382377 5 g 5g
11372377 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11382377 Supplier Thermo Scientific Alfa Aesar Supplier No. L09973.06
 Restricted Item
In Stock
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 2459-25-8 | C12H10O2 | 186.21 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 2459-25-8
Molecular Formula C12H10O2
Molecular Weight (g/mol) 186.21
MDL Number MFCD00017345
InChI Key IODOXLXFXNATGI-UHFFFAOYSA-N
Synonym methyl 2-naphthoate, methyl 2-naphthalenecarboxylate, 2-naphthalenecarboxylic acid, methyl ester, 2-naphthoic acid, methyl ester, 2-carbomethoxynaphthalene, 2-naphthalenecarboxylic acid methyl ester, acmc-1cliu, methyl-2-naphthoate, 2-naphthalenecarboxylicacid, methyl ester
PubChem CID 137605
IUPAC Name methyl naphthalene-2-carboxylate
SMILES COC(=O)C1=CC2=CC=CC=C2C=C1

Specifications

Melting Point 75°C to 79°C
Boiling Point 290°C
Quantity 5 g
Beilstein 2046448
Formula Weight 186.21
Percent Purity 99%
Chemical Name or Material Methyl 2-naphthoate

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.