missing translation for 'onlineSavingsMsg'
Learn More

methyl 2-(morpholinosulfonyl)benzoate, 95%, Thermo Scientific™

Product Code. 10172744
Click to view available options
Quantity:
1 g
5 g
Packaging:
Amber Glass Bottle
This item is not returnable. View return policy
This item is not returnable. View return policy

Chemical Identifiers

CAS 502182-56-1
Molecular Formula C12H15NO5S
Molecular Weight (g/mol) 285.31
MDL Number MFCD09879985
InChI Key FJLVVZZMDOHEEM-UHFFFAOYSA-N
Synonym methyl 2-morpholinosulfonyl benzoate, methyl 2-morpholine-4-sulfonyl benzoate, methyl 2-morpholin-4-yl sulphonyl benzoate
PubChem CID 341073
IUPAC Name methyl 2-morpholin-4-ylsulfonylbenzoate
SMILES COC(=O)C1=CC=CC=C1S(=O)(=O)N1CCOCC1

Specifications

CAS Min % 94.5
CAS Max % 100.0
Assay Percent Range 95%
Packaging Amber Glass Bottle
Quantity 1 g
Formula Weight 285.32
Percent Purity 95%
Chemical Name or Material Methyl 2-(morpholinosulfonyl)benzoate
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'