missing translation for 'onlineSavingsMsg'
Learn More

Methyl 2-methylthiopyrimidine-5-carboxylate, 97%, Thermo Scientific™

Product Code. 15229746 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
5 g
Unit Size:
5g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15229746 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 15229746 Supplier Thermo Scientific Acros Supplier No. 451440050

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Specifications

CAS 38275-41-1
Melting Point 96.0°C to 97.0°C
Infrared Spectrum Authentic
Assay Percent Range 96% min. (HPLC)
Molecular Formula C7H8N2O2S
MDL Number MFCD00194932
Quantity 5 g
Synonym methyl 2-methylthio pyrimidine-5-carboxylate, methyl 2-methylsulfanyl pyrimidine-5-carboxylate, methyl 2-methylthiopyrimidine-5-carboxylate, methyl 2-methylthio-5-pyrimidinecarboxylate, 2-methylthio-pyrimidine-5-carboxylic acid methyl ester, 5-pyrimidinecarboxylic acid, 2-methylthio-, methyl ester, methyl2-methylthio pyrimidine-5-carboxylate, methyl 2-methylthio-5-pyrimidinearboxylate, methyl-2-methylthio pyrimidine-5-carboxylate, methyl 2-methylsulphanyl pyrimidine-5-carboxylate
InChI Key PSYRMEZGAWNWHV-UHFFFAOYSA-N
SMILES COC(=O)C1=CN=C(SC)N=C1
IUPAC Name methyl 2-methylsulfanylpyrimidine-5-carboxylate
Molecular Weight (g/mol) 184.21
PubChem CID 11194721
Formula Weight 184.22
Percent Purity 97%
Chemical Name or Material Methyl 2-methylthiopyrimidine-5-carboxylate
Show More Show Less
compliance-icons
Product Identifier
  • Methyl 2-methylthiopyrimidine-5-carboxylate, 97%
Signal Word
  • Warning
Hazard Category
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H312-Harmful in contact with skin.
  • H332-Harmful if inhaled.
  • H302-Harmful if swallowed.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P312-Call a POISON CENTER/doctor/if you feel unwell.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.