missing translation for 'onlineSavingsMsg'
Learn More

Methyl 2-(methylthio)benzoate, 98%

Product Code. p-7081092
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
This item is not returnable. View return policy

Product Code. 11494273

Brand: Thermo Scientific Alfa Aesar A10905.03

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 3704-28-7 | C9H10O2S | 182.237 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 3704-28-7
Molecular Formula C9H10O2S
Molecular Weight (g/mol) 182.237
MDL Number MFCD00068039
InChI Key CPQDZXPLQXZJGF-UHFFFAOYSA-N
PubChem CID 279586
IUPAC Name methyl 2-methylsulfanylbenzoate
SMILES COC(=O)C1=CC=CC=C1SC

Specifications

Melting Point 64°C to 66°C
Odor Stench
Quantity 1 g
Beilstein 2084294
Synonym methyl 2-methylthio benzoate, methyl 2-methylsulfanyl benzoate, benzoic acid, 2-methylthio-, methyl ester, benzoic acid,2-methylthio-, methyl ester, 2-methylthiobenzoic acid, methyl ester, acmc-1cnec, methyl o-methylthiosalicylate, methyl-2-methylthio benzoate, methyl 2-methylsulfanyl benzoate #, 2-methylthio benzoic acid methyl ester
Formula Weight 182.24
Percent Purity 98%
Chemical Name or Material Methyl 2- (methylthio)benzoate

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.