missing translation for 'onlineSavingsMsg'
Learn More

Methyl 2-(methylthio)benzoate, 98%

Product Code. 11404283
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11404283 5 g 5g
11494273 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11404283 Supplier Thermo Scientific Alfa Aesar Supplier No. A10905.06
 Restricted Item
Estimated Shipment: 30-07-2026
Add to basket
This item is not returnable. View return policy

CAS: 3704-28-7 | C9H10O2S | 182.237 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 3704-28-7
Molecular Formula C9H10O2S
Molecular Weight (g/mol) 182.237
MDL Number MFCD00068039
InChI Key CPQDZXPLQXZJGF-UHFFFAOYSA-N
PubChem CID 279586
IUPAC Name methyl 2-methylsulfanylbenzoate
SMILES COC(=O)C1=CC=CC=C1SC

Specifications

Melting Point 64°C to 66°C
Odor Stench
Quantity 5 g
Beilstein 2084294
Synonym methyl 2-methylthio benzoate, methyl 2-methylsulfanyl benzoate, benzoic acid, 2-methylthio-, methyl ester, benzoic acid,2-methylthio-, methyl ester, 2-methylthiobenzoic acid, methyl ester, acmc-1cnec, methyl o-methylthiosalicylate, methyl-2-methylthio benzoate, methyl 2-methylsulfanyl benzoate #, 2-methylthio benzoic acid methyl ester
Formula Weight 182.24
Percent Purity 98%
Chemical Name or Material Methyl 2- (methylthio)benzoate

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.