missing translation for 'onlineSavingsMsg'
Learn More

Methyl 2-methylbutyrate, 98%, Thermo Scientific Chemicals

Catalog Number 15214814
Click to view available options
Quantity:
100 g
1 kg
5 kg
This item is not returnable. View return policy
868-57-5
C6H12O2
116.16
MFCD00009335
OCWLYWIFNDCWRZ-UHFFFAOYNA-N
methyl 2-methylbutyrate, butanoic acid, 2-methyl-, methyl ester, methyl 2-methyl butyrate, methyl s-2-methylbutanoate, 2-methylbutyric acid methyl ester, butyric acid, 2-methyl-, methyl ester, dl-2-methylbutyric acid methyl ester, methyl dl-2-methylbutyrate, methyl alpha-methylbutyrate, methyl alpha-methylbutanoate
13357
methyl 2-methylbutanoate
CCC(C)C(=O)OC
This item is not returnable. View return policy
868-57-5
C6H12O2
116.16
MFCD00009335
OCWLYWIFNDCWRZ-UHFFFAOYNA-N
methyl 2-methylbutyrate, butanoic acid, 2-methyl-, methyl ester, methyl 2-methyl butyrate, methyl s-2-methylbutanoate, 2-methylbutyric acid methyl ester, butyric acid, 2-methyl-, methyl ester, dl-2-methylbutyric acid methyl ester, methyl dl-2-methylbutyrate, methyl alpha-methylbutyrate, methyl alpha-methylbutanoate
13357
methyl 2-methylbutanoate
CCC(C)C(=O)OC

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Density 0.8850g/mL
Boiling Point 115.0°C
Flash Point 18°C
Assay Percent Range 97.5% min. (GC)
Packaging Glass bottle
Linear Formula CH3CH2CH(CH3)COOCH3
Quantity 100 g
Specific Gravity 0.885
Solubility Information Solubility in water: practically insoluble
Formula Weight 116.16
Percent Purity 98%
Chemical Name or Material Methyl 2-methylbutyrate
Show More Show Less
compliance-icons compliance-icons
Product Identifier
  • Methyl 2-methylbutyrate
Signal Word
  • Danger
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Flammable liquid Category 2
  • Skin corrosion/irritation Category 2
Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H225-Highly flammable liquid and vapour.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'