missing translation for 'onlineSavingsMsg'
Learn More

Methyl 2-methylbutyrate, 98%

Product Code. 15204814 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
100 g
1 kg
5 kg
Unit Size:
100g
1kg
5kg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15204814 1 kg 1kg
15214814 100 g 100g
15294804 5 kg 5kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 15204814 Supplier Thermo Scientific Acros Supplier No. 450230010
 Restricted Item
In Stock
Add to basket
This item is not returnable. View return policy

CAS: 868-57-5 | C6H12O2 | 116.16 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 868-57-5
Molecular Formula C6H12O2
Molecular Weight (g/mol) 116.16
MDL Number MFCD00009335
InChI Key OCWLYWIFNDCWRZ-UHFFFAOYNA-N
Synonym methyl 2-methylbutyrate, butanoic acid, 2-methyl-, methyl ester, methyl 2-methyl butyrate, methyl s-2-methylbutanoate, 2-methylbutyric acid methyl ester, butyric acid, 2-methyl-, methyl ester, dl-2-methylbutyric acid methyl ester, methyl dl-2-methylbutyrate, methyl alpha-methylbutyrate, methyl alpha-methylbutanoate
PubChem CID 13357
IUPAC Name methyl 2-methylbutanoate
SMILES CCC(C)C(=O)OC

Specifications

Density 0.8850g/mL
Boiling Point 115.0°C
Flash Point 18°C
Assay Percent Range 97.5% min. (GC)
Packaging Glass bottle
Linear Formula CH3CH2CH(CH3)COOCH3
Quantity 1 kg
Specific Gravity 0.885
Solubility Information Solubility in water: practically insoluble
Formula Weight 116.16
Percent Purity 98%
Chemical Name or Material Methyl 2-methylbutyrate
Show More Show Less
compliance-icons compliance-icons
Product Identifier
  • Methyl 2-methylbutyrate
Signal Word
  • Danger
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Flammable liquid Category 2
  • Skin corrosion/irritation Category 2
Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H225-Highly flammable liquid and vapour.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.