missing translation for 'onlineSavingsMsg'
Learn More

Methyl 2-(Methylamino)benzoate, TRC

Product Code. 30004224
Change view
Click to view available options
Quantity:
100 mg
500 mg
1 g
Unit Size:
100mg
1g
500mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30004224 1 g 1g
30007086 100 mg 100mg
30006995 500 mg 500mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30004224 Supplier Toronto Research Chemicals Supplier No. TRCM2919761G
 Restricted Item
Estimated Shipment: 25-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 85-91-6
Molecular Formula C9H11NO2
Molecular Weight (g/mol) 165.19
Synonym 2-(Methylamino)benzoic acid methyl ester, Methyl N-methylanthranilate, Methyl methanthranilate, Methyl methylanthranilate, Methyl o-(methylamino)benzoate, N-Methylanthranilic acid methyl ester, NSC 9406
IUPAC Name methyl 2-(methylamino)benzoate
SMILES CNc1ccccc1C(=O)OC

Specifications

Quantity 1 g
InChI Formula InChI=1S/C9H11NO2/c1-10-8-6-4-3-5-7(8)9(11)12-2/h3-6,10H,1-2H3
Formula Weight 165.079
Chemical Name or Material Methyl 2-(Methylamino)benzoate
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.