missing translation for 'onlineSavingsMsg'
Learn More

Methyl 2-methyl-3-nitrobenzoate, 97%

Product Code. 15446718
Change view
Click to view available options
Quantity:
5 g
25 g
Unit Size:
25g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15446718 5 g 5g
15456718 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15446718 Supplier Thermo Scientific Alfa Aesar Supplier No. H60242.06
 Restricted Item
Estimated Shipment: 27-08-2026
Add to basket
This item is not returnable. View return policy

CAS: 59382-59-1 | C9H9NO4 | 195.17 g/mol

Methyl 2-methyl-3-nitrobenzoate may be used in the synthesis of methyl indole-4-carboxylate, 5-aminoisoquinolin-1(2H)-one, 5-nitroisocoumarin and [2-(4-fluorophenyl)-1H-indol-4-yl]-1-pyrrolidinylmethanone. It may also be used in the preparation of substituted nitrostyrene benzoic acids, via reaction with aromatic aldehydes in the presence of DBU in DMSO; and 4-(hydroxymethyl)-1-tosylindole, via Batcho-Leimgruber modification of the Reissert indole synthesis.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Methyl 2-methyl-3-nitrobenzoate may be used in the synthesis of methyl indole-4-carboxylate, 5-aminoisoquinolin-1(2H)-one, 5-nitroisocoumarin and [2-(4-fluorophenyl)-1H-indol-4-yl]-1-pyrrolidinylmethanone. It may also be used in the preparation of substituted nitrostyrene benzoic acids, via reaction with aromatic aldehydes in the presence of DBU in DMSO; and 4-(hydroxymethyl)-1-tosylindole, via Batcho-Leimgruber modification of the Reissert indole synthesis.

Solubility
Soluble in methanol.

Notes
Stable under recommended storage conditions. Incompatible with oxidizing agents and heat.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 59382-59-1
Molecular Formula C9H9NO4
Molecular Weight (g/mol) 195.17
MDL Number MFCD00082564
InChI Key CRZGFIMLHZTLGT-UHFFFAOYSA-N
Synonym benzoic acid, 2-methyl-3-nitro-, methyl ester, methyl 3-nitro-o-toluate, 2-methyl-3-nitrobenzoic acid methyl ester, methyl 3-nitro-2-methylbenzoate, 3-nitro-o-toluic acid methyl ester, 2-methyl-3-nitromethylbenzoate, pubchem10933, acmc-1awh8, ksc495s1j, methyl2-methyl-3-nitrobenzoate
PubChem CID 601165
IUPAC Name methyl 2-methyl-3-nitrobenzoate
SMILES COC(=O)C1=CC=CC(=C1C)[N+]([O-])=O

Specifications

Melting Point 62°C to 65°C
Quantity 5 g
Beilstein 1964020
Solubility Information Soluble in methanol.
Formula Weight 195.17
Percent Purity 97%
Chemical Name or Material Methyl 2-methyl-3-nitrobenzoate

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.