missing translation for 'onlineSavingsMsg'
Learn More

Methyl 2-methyl-3-furoate, 97%

Product Code. 11484004
Change view
Click to view available options
Quantity:
25 g
100 g
Unit Size:
100g
25g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11484004 100 g 100g
11474004 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11484004 Supplier Thermo Scientific Alfa Aesar Supplier No. A11979.22
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 6141-58-8 | C7H8O3 | 140.138 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 6141-58-8
Molecular Formula C7H8O3
Molecular Weight (g/mol) 140.138
MDL Number MFCD00003249
InChI Key UVRRIABXNIGUJZ-UHFFFAOYSA-N
PubChem CID 80237
IUPAC Name methyl 2-methylfuran-3-carboxylate
SMILES CC1=C(C=CO1)C(=O)OC

Specifications

Density 1.114
Boiling Point 71°C to 72°C (20 mmHg)
Flash Point 63°C (145°F)
Refractive Index 1.4745
Quantity 100 g
Beilstein 1342040
Synonym methyl 2-methyl-3-furoate, methyl 2-methyl-3-furancarboxylate, 2-methyl-furan-3-carboxylic acid methyl ester, 3-furancarboxylic acid, 2-methyl-, methyl ester, 3-furoic acid, 2-methyl-, methyl ester, methyl2-methyl-3-furoate, acmc-1bcvo, methyl 2-methyl-3-furoate #, methyl-2-methyl-3-furoate, 2-methylfuran-3-carboxylic acid methyl
Formula Weight 140.14
Percent Purity 97%
Chemical Name or Material Methyl 2-methyl-3-furoate

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.