missing translation for 'onlineSavingsMsg'
Learn More

Methyl 2-methoxypropionate, 98%

Product Code. 11393117
Change view
Click to view available options
Quantity:
5 g
25 g
Unit Size:
25g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11393117 5 g 5g
11303127 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11393117 Supplier Thermo Scientific Alfa Aesar Supplier No. L13527.06
 Restricted Item
In Stock
Add to basket
This item is not returnable. View return policy

CAS: 17639-76-8 | C5H10O3 | 118.132 g/mol

Methyl 2-methoxypropionate, is used as a pharmaceutical intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Methyl 2-methoxypropionate, is used as a pharmaceutical intermediate.

Solubility
Soluble in water.

Notes
Keep container tightly closed in a dry and well-ventilated place. Keep away from oxidizing agents. Stable under recommended storage conditions.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 17639-76-8
Molecular Formula C5H10O3
Molecular Weight (g/mol) 118.132
MDL Number MFCD00151664
InChI Key VABBJJOSOCPYIT-UHFFFAOYSA-N
Synonym methyl 2-methoxypropionate, propanoic acid, 2-methoxy-, methyl ester, acmc-20akie, d +-alpha-methoxy-propionic acid methyl ester, methyl2-methoxypropanoate, methyl 2-methoxy-propionate, 2-methoxypropanoic acid methyl ester, racemic 2-methoxy-propionic acid methyl ester
PubChem CID 86599
IUPAC Name methyl 2-methoxypropanoate
SMILES CC(C(=O)OC)OC

Specifications

Boiling Point 43°C to 44°C (15 mmHg)
Flash Point 36°C (96°F)
Refractive Index 1.397
Quantity 5 g
UN Number UN3272
Beilstein 1720819
Solubility Information Soluble in water.
Formula Weight 118.13
Percent Purity 98%
Chemical Name or Material Methyl 2-methoxypropionate

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.