missing translation for 'onlineSavingsMsg'
Learn More

Methyl 2-hydroxynicotinate, 97%

Product Code. 15496137
Change view
Click to view available options
Quantity:
250 mg
1 g
5 g
Unit Size:
1g
250mg
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15496137 5 g 5g
15406147 250 mg 250mg
15486137 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 15496137 Supplier Thermo Scientific Alfa Aesar Supplier No. H33334.06
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 10128-91-3 | C7H7NO3 | 153.137 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 10128-91-3
Molecular Formula C7H7NO3
Molecular Weight (g/mol) 153.137
MDL Number MFCD00661282
InChI Key SILBTMNCGYLTOK-UHFFFAOYSA-N
Synonym methyl 2-hydroxynicotinate, methyl 2-oxo-1,2-dihydro-3-pyridinecarboxylate, methyl 2-oxo-1,2-dihydropyridine-3-carboxylate, methyl 2-hydroxypyridine-3-carboxylate, 2-hydroxy-nicotinic acid methyl ester, 2-hydroxynicotinic acid methyl ester, 2-hydroxy-3-pyridinecarboxylic acid methyl ester, 3-pyridinecarboxylic acid, 2-hydroxy-, methyl ester, methyl 2-hydroxy-3-pyridinecarboxylate, 3-pyridinecarboxylic acid, 1,2-dihydro-2-oxo-, methyl ester
PubChem CID 579028
IUPAC Name methyl 2-oxo-1H-pyridine-3-carboxylate
SMILES COC(=O)C1=CC=CNC1=O

Specifications

Melting Point 153°C to 155°C
Color Cream
Quantity 5 g
Formula Weight 153.1
Percent Purity 97%
Physical Form Solid
Chemical Name or Material Methyl 2-hydroxynicotinate

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.