missing translation for 'onlineSavingsMsg'
Learn More

Methyl 2-hydroxyisobutyrate, 99%

Product Code. 10367611 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
5 g
50 g
Packaging:
Glass bottle
Unit Size:
50g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10367611 5 g Glass bottle 5g
10631642 50 g Glass bottle 50g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10367611 Supplier Thermo Scientific Acros Supplier No. 126970050
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 2110-78-3 | C5H10O3 | 118.13 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 2110-78-3
Molecular Formula C5H10O3
Molecular Weight (g/mol) 118.13
MDL Number MFCD00004457
InChI Key XYVQFUJDGOBPQI-UHFFFAOYSA-N
Synonym methyl 2-hydroxyisobutyrate, methyl 2-methyllactate, 2-hydroxyisobutyric acid methyl ester, methyl 2-hydroxy-2-methylpropionate, methyl2-hydroxyisobutyrate, propanoic acid, 2-hydroxy-2-methyl-, methyl ester, methyl alpha-hydroxyisobutyrate, methyl .alpha.-hydroxyisobutyrate, methyl ester of 2-methyllactic acid, methyl 2-hydroxy-2-methyl-propanoate
PubChem CID 75024
IUPAC Name methyl 2-hydroxy-2-methylpropanoate
SMILES CC(C)(C(=O)OC)O

Specifications

Color Colorless
Density 1.0300g/mL
Boiling Point 137°C
Flash Point 41°C
Infrared Spectrum Authentic
Assay Percent Range 98.5% min. (GC)
Packaging Glass bottle
Linear Formula (CH3)2C(OH)CO2CH3
Refractive Index 1.4100 to 1.412
Quantity 5 g
Beilstein 03, II, 223
Specific Gravity 1.03
Formula Weight 118.13
Percent Purity 99%
Physical Form Liquid
Chemical Name or Material Methyl 2-hydroxyisobutyrate, 99%
Show More Show Less
compliance-icons
Product Identifier
  • Methyl 2-hydroxyisobutyrate
Signal Word
  • Warning
Hazard Category
  • Flammable liquid Category 3
Hazard Statement
  • H226-Flammable liquid and vapour.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.