missing translation for 'onlineSavingsMsg'
Learn More

Methyl 2-hydroxy-5-nitrobenzoate, 98%

Product Code. 11490848
Change view
Click to view available options
Quantity:
10 g
50 g
250 g
Unit Size:
10g
250g
50g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11490848 250 g 250g
11470848 10 g 10g
11480848 50 g 50g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11490848 Supplier Thermo Scientific Alfa Aesar Supplier No. B25278.30
 Restricted Item
Estimated Shipment: 11-08-2026
Add to basket
This item is not returnable. View return policy

CAS: 17302-46-4 | C8H7NO5 | 197.146 g/mol

Methyl 2-hydroxy-5-nitrobenzoate is used as pharmaceutical intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Methyl 2-hydroxy-5-nitrobenzoate is used as pharmaceutical intermediate.

Solubility
Insoluble in water.

Notes
Stable under recommended storage conditions. Incompatible with oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 17302-46-4
Molecular Formula C8H7NO5
Molecular Weight (g/mol) 197.146
MDL Number MFCD00024695
InChI Key UUBFELFUKFJSRD-UHFFFAOYSA-N
Synonym methyl 2-hydroxy-5-nitrobenzoate, methyl 5-nitrosalicylate, methyl-5-nitrosalicylate, 5-nitrosalicylic acid methyl ester, benzoic acid, 2-hydroxy-5-nitro-, methyl ester, 2-hydroxy-5-nitrobenzoic acid methyl ester, methyl 5-nitro-salicylate, acmc-20a62q, methyl 5-nitro-2-hydroxybenzoate, methyl-2-hydroxy-5-nitrobenzoate
PubChem CID 87041
SMILES COC(=O)C1=C(C=CC(=C1)[N+](=O)[O-])O

Specifications

Melting Point 114°C to 117°C
Quantity 250 g
Solubility Information Insoluble in water.
Formula Weight 197.15
Percent Purity 98%
Chemical Name or Material Methyl 2-hydroxy-5-nitrobenzoate

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.