missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Methyl 2-hydroxy-3-nitrobenzoate, 98%
Click to view available options
Quantity:
5 g
25 g
Unit Size:
25g
5g
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Specifications
| Melting Point | 129°C to 132°C |
| Quantity | 5 g |
| Formula Weight | 197.15 |
| Percent Purity | 98% |
| Chemical Name or Material | Methyl 2-hydroxy-3-nitrobenzoate |
RUO – Research Use Only
Berichtigung von Produktinhalten
Bitte geben Sie uns Ihr Feedback zu den Produktinhalten, indem Sie das folgende Formular ausfüllen.
Name des Produkts
Haben Sie Verbesserungsvorschläge?Übermitteln Sie eine inhaltliche Korrektur