missing translation for 'onlineSavingsMsg'
Learn More

Methyl 2-hexynoate, 98+%, Thermo Scientific™

Product Code. 11466256
Click to view available options
Quantity:
5 g
25 g
Unit Size:
25g
5g
This item is not returnable. View return policy

Product Code. 11466256

Brand: Thermo Scientific Alfa Aesar B23065.14

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 18937-79-6
Molecular Formula C7H10O2
Molecular Weight (g/mol) 126.155
MDL Number MFCD00041642
InChI Key DHZPZBFOJDXEHS-UHFFFAOYSA-N
Synonym methyl 2-hexynoate, 2-hexynoic acid methyl ester, 2-hexynoic acid, methyl ester, hex-2-ynoic acid methyl ester, methyl hexynoate, acmc-20akik, 2-hexynoicacid,methyl ester, 2-hexynoicacid, methyl ester
PubChem CID 87860
IUPAC Name methyl hex-2-ynoate
SMILES CCCC#CC(=O)OC

Specifications

Density 0.944
Boiling Point 64°C to 66°C (10 mmHg)
Flash Point 67°C (152°F)
Refractive Index 1.442
Quantity 25 g
Beilstein 1750018
Formula Weight 126.16
Percent Purity ≥98%
Chemical Name or Material Methyl 2-hexynoate
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.