missing translation for 'onlineSavingsMsg'
Learn More

Methyl 2-furoate, MedChemExpress

Product Code. 16455467
Encompass_Preferred
Change view
Click to view available options
Quantity:
50 mg
Unit Size:
50mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16455467 50 mg 50mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 16455467 Supplier Medchem Express Supplier No. HYY0949
 Restricted Item

This item requires a price update prior to being ordered.

Request a Price for this item here.

This item is not returnable. View return policy

MedChemExpress Methyl 2-furoate (Methyl furan-2-carboxylate) is a building block in chemical synthesis. A flavoring agent in food. Found in cranberries, guava fruits, raisins and other fruits. Also present in baked potato, roasted filberts, roasted peanut, tomatoes, coffee, cocoa, okra, etc.

Methyl 2-furoate (Methyl furan-2-carboxylate) is a building block in chemical synthesis. A flavoring agent in food. Found in cranberries, guava fruits, raisins and other fruits. Also present in baked potato, roasted filberts, roasted peanut, tomatoes, coffee, cocoa, okra, etc.

Specifications

Chemical Name or Material Methyl 2-furoate
CAS 611-13-2
Color Colorless
Purity Grade Notes Research
Molecular Formula C6H6O3
Quantity 50 mg
Synonym Methyl furan-2-carboxylate
Solubility Information DMSO : 250 mg/mL (1982.40 mM; Need ultrasonic)
SMILES O=C(C1=CC=CO1)OC
Molecular Weight (g/mol) 126.11
Formula Weight 126.11
Percent Purity 99.75%
Grade Research
Physical Form Liquid
For Use With (Application) Metabolism-protein/nucleotide metabolism
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.