missing translation for 'onlineSavingsMsg'
Learn More

Methyl (2-furfurylthio)acetate, 97%, Thermo Scientific™

Product Code. 11465367
Click to view available options
Quantity:
5 g
25 g
100 g
Unit Size:
100g
25g
5g
This item is not returnable. View return policy

Product Code. 11465367

Brand: Thermo Scientific Alfa Aesar B22717.06

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 108499-33-8
Molecular Formula C8H10O3S
Molecular Weight (g/mol) 186.23
MDL Number MFCD02283460
InChI Key XPVNCXPHDABNLE-UHFFFAOYSA-N
Synonym methyl 2-furfurylthio acetate, methyl 2-furan-2-ylmethyl thio acetate, acetic acid,2-2-furanylmethyl thio-, methyl ester, methyl 2-furan-2-ylmethyl sulfanyl acetate, acmc-20f6v3, aceticacid, 2-furanylmethyl thio-methylester, methyl-2-furfurylthio acetate, methyl 2-2-furylmethylthio acetate, furfurylthio acetic acid methyl ester, 2-furfurylthio acetic acid methyl ester
PubChem CID 7022533
IUPAC Name methyl 2-(furan-2-ylmethylsulfanyl)acetate
SMILES COC(=O)CSCC1=CC=CO1

Specifications

Boiling Point 95°C (3 mmHg)
Flash Point >100°C (212°F)
Quantity 5 g
Formula Weight 186.23
Percent Purity 97%
Chemical Name or Material Methyl (2-furfurylthio)acetate
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.