missing translation for 'onlineSavingsMsg'
Learn More

Methyl 2-formylthiophene-4-carboxylate, 95%

Product Code. 11476510
Change view
Click to view available options
Quantity:
250 mg
1 g
5 g
Unit Size:
1g
250mg
5g
Product Code. Quantity unitSize
11476510 5 g 5g
15463727 250 mg 250mg
11466510 1 g 1g
3 options
This item is not returnable. View return policy

Product Code. 11476510

Brand: Thermo Scientific Alfa Aesar H27597.06

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

CAS: 67808-66-6 | C7H6O3S | 170.182 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 67808-66-6
Molecular Formula C7H6O3S
Molecular Weight (g/mol) 170.182
MDL Number MFCD01859945
InChI Key GNXNZRYWBFMVHK-UHFFFAOYSA-N
Synonym methyl 2-formyl-4-thiophenecarboxylate, 2-formylthiophene-4-carboxylic acid methylester, 5-formyl-3-thiophenecarboxylic acid methyl ester, 5-formyl-3-thiophene carboxylic acid methylester, pubchem7741, tpc-h014, methyl 5-methanoylthiophene-3-carboxylate, 5-formyl-thiophene-3-carboxylic acid methyl ester, 3-thiophenecarboxylic acid, 5-formyl-, methyl ester, 3-thiophenecarboxylicacid, 5-formyl-, methyl ester
PubChem CID 818925
IUPAC Name methyl 5-formylthiophene-3-carboxylate
SMILES COC(=O)C1=CSC(=C1)C=O

Specifications

Melting Point 113°C to 117°C
Quantity 5 g
Sensitivity Air Sensitive
Formula Weight 170.19
Percent Purity 95%
Chemical Name or Material Methyl 2-formylthiophene-4-carboxylate

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.