missing translation for 'onlineSavingsMsg'
Learn More

Methyl 2-formylbenzoate, Thermo Scientific™, 97%, Thermo Scientific™

Product Code. 10089441
Change view
Click to view available options
Quantity:
1 g
10 g
50 g
Unit Size:
10g
1g
50g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
10089441 50 g 50g
10541323 1 g 1g
10672073 10 g 10g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 10089441 Supplier Thermo Scientific Maybridge Supplier No. TL00698EE
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Chemical Identifiers

CAS 4122-56-9
Molecular Formula C9H8O3
Molecular Weight (g/mol) 164.16
MDL Number MFCD00204231
InChI Key YRMODRRGEUGHTF-UHFFFAOYSA-N
Synonym 2-carbomethoxybenzaldehyde, methyl o-formylbenzoate, phthalaldehydic acid, methyl ester, benzoic acid, formyl-, methyl ester, methyl 2-formylbenzenecarboxylate, benzoic acid, 2-formyl-, methyl ester, methyl 2-formyl-benzoate, methylformylbenzenecarboxylate, methyl-2-formylbenzoate, formyl-benzoic acid methyl ester
PubChem CID 243003
IUPAC Name methyl 2-formylbenzoate
SMILES COC(=O)C1=CC=CC=C1C=O

Specifications

CAS Min % 97.0
Assay Percent Range 97%
Linear Formula CH3OC(O)C6H4CHO
Quantity 50 g
Formula Weight 164.16
Percent Purity 97%
Chemical Name or Material Methyl 2-formylbenzoate
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.