missing translation for 'onlineSavingsMsg'
Learn More

Methyl 2-fluorophenylacetate, 99%, Thermo Scientific™

Product Code. 10780961 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
Packaging:
Glass bottle
Unit Size:
100g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10780961 5 g Glass bottle 5g
10247180 25 g Glass bottle 25g
11904921 100 g Glass bottle 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 10780961 Supplier Thermo Scientific Acros Supplier No. 304640050
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Chemical Identifiers

CAS 57486-67-6
Molecular Formula C9H9FO2
Molecular Weight (g/mol) 168.17
MDL Number MFCD00800605
InChI Key TWXVEZBPKFIMBB-UHFFFAOYSA-N
Synonym methyl 2-2-fluorophenyl acetate, methyl 2-fluorophenyl acetate, methyl 2-fluorophenylacetate, 2-fluorophenylacetic acid methyl ester, benzeneacetic acid, 2-fluoro-, methyl ester, 2-fluoro-phenyl-acetic acid methyl ester, methyl 2-2-fluorophenyl ethanoate, 2-fluoro-phenylacetic acid methyl ester, 2-2-fluorophenyl acetic acid methyl ester
PubChem CID 2733230
IUPAC Name methyl 2-(2-fluorophenyl)acetate
SMILES COC(=O)CC1=CC=CC=C1F

Specifications

Color Colorless
Infrared Spectrum Authentic
Assay Percent Range 98.5% min. (GC)
Packaging Glass bottle
Refractive Index 1.4890 to 1.4910
Quantity 5 g
Formula Weight 168.06
Percent Purity 99%
Physical Form Liquid
Chemical Name or Material Methyl 2-fluorophenylacetate

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.