missing translation for 'onlineSavingsMsg'
Learn More

Methyl 2-fluorobenzoate, 98%

Product Code. 11453177
Click to view available options
Quantity:
5 g
25 g
Unit Size:
25g
5g
This item is not returnable. View return policy

Product Code. 11453177

Brand: Thermo Scientific Alfa Aesar A16133.06

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 394-35-4 | C8H7FO2 | 154.14 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 394-35-4
Molecular Formula C8H7FO2
Molecular Weight (g/mol) 154.14
MDL Number MFCD00017913
InChI Key QAFJIJWLEBLXHH-UHFFFAOYSA-N
Synonym methyl o-fluorobenzoate, methyl-2-fluorobenzoate, 2-fluorobenzoic acid methyl ester, benzoic acid, 2-fluoro-, methyl ester, o-fluorobenzoic acid, methyl ester, benzoic acid, o-fluoro-, methyl ester, 2-fluoro-benzoic acid methyl ester, methyl2-fluorobenzoate, pubchem3565, acmc-209j5e
PubChem CID 67854
IUPAC Name methyl 2-fluorobenzoate
SMILES COC(=O)C1=CC=CC=C1F

Specifications

Density 1.21
Boiling Point 109°C to 110°C (35 mmHg)
Flash Point 93°C (199°F)
Odor Aromatic
Refractive Index 1.503
Quantity 5 g
Beilstein 1862493
Formula Weight 154.14
Percent Purity 98%
Chemical Name or Material Methyl 2-fluorobenzoate
compliance-icons
Product Identifier
  • Methyl 2-fluorobenzoate
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/soap.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P312-Call a POISON CENTER/doctor/if you feel unwell.
  • P332+P313-If skin irritation occurs: Get medical advice/attention.
  • P337+P313-If eye irritation persists: Get medical advice/attention.
Supplemental information
  • MIXTURE LIST-Contains : Methyl 2-fluorobenzoate

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.