missing translation for 'onlineSavingsMsg'
Learn More

Methyl 2-fluoro-5-nitrobenzoate, 98%

Product Code. 15488238
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
Unit Size:
100g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15488238 5 g 5g
15498238 25 g 25g
15408248 100 g 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 15488238 Supplier Thermo Scientific Alfa Aesar Supplier No. H61676.06
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 2965-22-2 | C8H6FNO4 | 199.137 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 2965-22-2
Molecular Formula C8H6FNO4
Molecular Weight (g/mol) 199.137
MDL Number MFCD06203689
InChI Key JZLONOOYIXEAHM-UHFFFAOYSA-N
Synonym methyl 2-fluoro-5-nitrobenzenecarboxylate, 2-fluoro-5-nitrobenzoic acid methyl ester, 5-nitro-2-fluorobenzoic acid methyl ester, benzoicacid, 2-fluoro-5-nitro-, methyl ester, benzoic acid, 2-fluoro-5-nitro-, methyl ester, methyl 2-fluoro-5-nitro-benzoate, acmc-209ha6, ksc563m1n, methyl2-fluoro-5-nitrobenzoate, 4-fluoro-3-methoxycarbonyl nitrobenzene
PubChem CID 11217860
IUPAC Name methyl 2-fluoro-5-nitrobenzoate
SMILES COC(=O)C1=C(C=CC(=C1)[N+](=O)[O-])F

Specifications

Quantity 5 g
Formula Weight 199.14
Percent Purity 98%
Physical Form Solid
Chemical Name or Material Methyl 2-fluoro-5-nitrobenzoate

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.