missing translation for 'onlineSavingsMsg'
Learn More

Methyl 2-fluoro-4-hydroxybenzoate, 98%

Código de producto. 15458238
Cambiar vista
Click to view available options
Quantity:
250 mg
1 g
5 g
Tamaño de la unidad:
1g
250mg
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Código de producto. Quantity Tamaño de la unidad
15458238 1 g 1g
15478238 250 mg 250mg
15468238 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Este artículo no se puede devolver. Vea la política de devoluciones
Código de producto. 15458238 Proveedor Thermo Scientific Alfa Aesar N.º de proveedor H61675.03
 Restricted Item
Estimated Shipment: 21-10-2026
Añadir a la cesta
Añadir a la cesta
Request Bulk or Custom Formats

missing translation for 'validMainframeMsg'

Este artículo no se puede devolver. Vea la política de devoluciones

CAS: 197507-22-5 | C8H7FO3 | 170.139 g/mol

Methyl 2-fluoro-4-hydroxybenzoate can be used in the preparation of compounds, with the ability to inhibit cytosolic phospholipase A2α. It is also an intermediate in the synthesis of 3-(2-fluoro-4-hydroxyphenyl)-5-(trifluoromethyl) -4-isoxazolecarboxylic acid methyl ester (F591865).

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Methyl 2-fluoro-4-hydroxybenzoate can be used in the preparation of compounds, with the ability to inhibit cytosolic phospholipase A2α. It is also an intermediate in the synthesis of 3-(2-fluoro-4-hydroxyphenyl)-5-(trifluoromethyl) -4-isoxazolecarboxylic acid methyl ester (F591865).

Solubility
Slightly soluble in water.

Notes
Stable under recommended storage conditions. Incompatible with oxidizing agents.
TRUSTED_SUSTAINABILITY

Identificadores de productos químicos

CAS 197507-22-5
Molecular Formula C8H7FO3
Molecular Weight (g/mol) 170.139
MDL Number MFCD09038282
InChI Key WYSPMXSNCAFCFV-UHFFFAOYSA-N
Synonym 2-fluoro-4-hydroxy-benzoic acid methyl ester, 2-fluoro-4-hydroxybenzoic acid methyl ester, benzoic acid, 2-fluoro-4-hydroxy-, methyl ester, methyl-2-fluoro4-hydroxybenzoate, methyl2-fluoro-4-hydroxybenzoate, methyl-2-fluoro-4-hydroxybenzoate, methyl 2-fluoro-4-hydroxy-benzoate, 2-fluoro-4-hyroxybenzoic acid methyl ester
PubChem CID 22463192
IUPAC Name methyl 2-fluoro-4-hydroxybenzoate
SMILES COC(=O)C1=C(C=C(C=C1)O)F

Especificaciones

Quantity 1 g
Solubility Information Slightly soluble in water.
Formula Weight 170.14
Percent Purity 98%
Chemical Name or Material Methyl 2-fluoro-4-hydroxybenzoate

RUO – Research Use Only

Título del producto
Seleccione un problema

Al hacer clic en Enviar, acepta que Fisher Scientific se ponga en contacto con usted en relación con los comentarios que ha proporcionado en este formulario. No compartiremos su información para ningún otro fin. Toda la información de contacto proporcionada se mantendrá de acuerdo con nuestra Política de Privacidad. Política de privacidad.