missing translation for 'onlineSavingsMsg'
Learn More

Methyl 2-(chlorosulfonyl)benzoate, 94%

Product Code. p-7026904
Click to view available options
Quantity:
5 g
25 g
100 g
Unit Size:
100g
25g
5g
This item is not returnable. View return policy

Product Code. 11400817

Brand: Thermo Scientific Alfa Aesar B23211.06

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 26638-43-7 | C8H7ClO4S | 234.65 g/mol

Methyl 2-(chlorosulfonyl)benzoate can be used as intermediates in pharmaceuticals, saccharin, dyes, & pigments.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Methyl 2-(chlorosulfonyl)benzoate can be used as intermediates in pharmaceuticals, saccharin, dyes, & pigments.

Solubility
Reacts with water.

Notes
Moisture sensitive. Store under inert gas. Store away from oxidizing agents, water/ moisture.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 26638-43-7
Molecular Formula C8H7ClO4S
Molecular Weight (g/mol) 234.65
MDL Number MFCD00009797
InChI Key HUNUAFNLLYVTQD-UHFFFAOYSA-N
PubChem CID 117830
SMILES COC(=O)C1=CC=CC=C1S(Cl)(=O)=O

Specifications

Melting Point 59°C to 64°C
Quantity 5 g
UN Number UN3261
Sensitivity Moisture sensitive
Synonym methyl 2-chlorosulfonyl benzoate, benzoic acid, 2-chlorosulfonyl-, methyl ester, unii-c6hzm8x6pk, c6hzm8x6pk, methyl 2-chlorosulphonyl benzoate, pubchem14808, acmc-1cirj, methyl 2-chiorosulfonylbenzoate, methyl 2 chlorosulfonyl benzoate, methyl2-chlorosulfonyl benzoate
Solubility Information Reacts with water.
IUPAC Name methyl 2-chlorosulfonylbenzoate
Formula Weight 234.66
Percent Purity 94%
Chemical Name or Material Methyl 2-(chlorosulfonyl)benzoate

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.