missing translation for 'onlineSavingsMsg'
Learn More

Methyl 2-Chloropropionate, TRC

Product Code. 30477342
Change view
Click to view available options
Quantity:
1 g
Unit Size:
1g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30477342 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30477342 Supplier Toronto Research Chemicals Supplier No. TRCM2971051G

Please to purchase this item. Need a web account? Register with us today!

This item requires a price update prior to being ordered.

Request a Price for this item here.

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Methyl 2-Chloropropionate
CAS 17639-93-9
Molecular Formula C4H7ClO2
Quantity 1 g
Synonym Methyl 2-Chloropropanoate
InChI Formula InChI=1S/C4H7ClO2/c1-3(5)4(6)7-2/h3H,1-2H3
SMILES COC(=O)C(C)Cl
IUPAC Name methyl 2-chloropropanoate
Molecular Weight (g/mol) 122.55
Formula Weight 122.0135
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.