missing translation for 'onlineSavingsMsg'
Learn More

Methyl 2-chlorobenzoate, 98%

Product Code. 11423574
Change view
Click to view available options
Quantity:
25 g
100 g
500 g
Unit Size:
100g
25g
500g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11423574 500 g 500g
11403574 25 g 25g
11413574 100 g 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11423574 Supplier Thermo Scientific Alfa Aesar Supplier No. A11237.36
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 610-96-8 | C8H7ClO2 | 170.59 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 610-96-8
Molecular Formula C8H7ClO2
Molecular Weight (g/mol) 170.59
MDL Number MFCD00016337
InChI Key JAVRNIFMYIJXIE-UHFFFAOYSA-N
Synonym methyl o-chlorobenzoate, methyl-2-chlorobenzoate, benzoic acid, 2-chloro-, methyl ester, 2-chlorobenzoic acid methyl ester, unii-g344nes07r, benzoic acid, o-chloro-, methyl ester, benzoicacid, 2-chloro-, methyl ester, pubchem3707, methyl-o-chlorobenzoate, acmc-209moz
PubChem CID 11895
IUPAC Name methyl 2-chlorobenzoate
SMILES COC(=O)C1=CC=CC=C1Cl

Specifications

Density 1.191
Boiling Point 233°C to 235°C
Flash Point 108°C (226°F)
Refractive Index 1.536
Quantity 500 g
Beilstein 1364684
Formula Weight 170.6
Percent Purity 98%
Chemical Name or Material Methyl 2-chlorobenzoate

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.