missing translation for 'onlineSavingsMsg'
Learn More

Methyl 2-chloroacetoacetate, 95%

Product Code. 10129232 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
10 mL
50 mL
Packaging:
Glass bottle
Unit Size:
10mL
50mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10129232 10 mL Glass bottle 10mL
10509643 50 mL Glass bottle 50mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10129232 Supplier Thermo Scientific Acros Supplier No. 179900100
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 4755-81-1 | C5H7ClO3 | 150.56 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 4755-81-1
Molecular Formula C5H7ClO3
Molecular Weight (g/mol) 150.56
MDL Number MFCD00008757
InChI Key GYQRIAVRKLRQKP-UHFFFAOYSA-N
Synonym methyl 2-chloroacetoacetate, butanoic acid, 2-chloro-3-oxo-, methyl ester, 2-chloroacetoacetic acid methyl ester, 2-chloro-3-oxo-butyric acid methyl ester, methyl2-chloroacetoacetate, acmc-209k9v, methyl-2-chloroacetoacetate, methyl-alpha-chloroacetoacetate, 2-chloroacetoacetate methyl ester, gyqriavrklrqkp-uhfffaoysa
PubChem CID 107332
IUPAC Name methyl 2-chloro-3-oxobutanoate
SMILES CC(=O)C(C(=O)OC)Cl

Specifications

Melting Point -33°C
Color Yellow
Density 1.2400g/mL
Boiling Point 137°C
Flash Point 72°C
Infrared Spectrum Authentic
Assay Percent Range 95%
Packaging Glass bottle
Linear Formula CH3COCH(Cl)CO2CH3
Refractive Index 1.4455 to 1.4475
Quantity 10 mL
Specific Gravity 1.24
Solubility Information Solubility in water: 50g/L (20°C). Other solubilities: soluble in organic solvents
Formula Weight 150.56
Percent Purity 95%
Physical Form Liquid
Chemical Name or Material Methyl 2-chloroacetoacetate
Show More Show Less
compliance-icons
Product Identifier
  • Methyl 2-chloroacetoacetate
Signal Word
  • Warning
Hazard Category
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H302-Harmful if swallowed.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.