missing translation for 'onlineSavingsMsg'
Learn More

Methyl 2-chloro-4-(pentafluoroethyl)nicotinate, 96%, Thermo Scientific™

Product Code. 15490087
Change view
Click to view available options
Quantity:
100 mg
Unit Size:
100mg
Product Code. Quantity unitSize
15490087 100 mg 100mg
1 options
This item is not returnable. View return policy

Product Code. 15490087

Brand: Thermo Scientific Alfa Aesar H66173.MC

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Specifications

CAS 1580464-71-6
Color Yellow
Molecular Formula C9H5ClF5NO2
Quantity 100 mg
Synonym Methyl 2-chloro-4-(pentafluoroethyl)pyridine-3-carboxylate
InChI Key GUORMFCKJWQHIU-UHFFFAOYSA-N
SMILES COC(=O)C1=C(C=CN=C1Cl)C(C(F)(F)F)(F)F
IUPAC Name methyl 2-chloro-4-(1,1,2,2,2-pentafluoroethyl)pyridine-3-carboxylate
Molecular Weight (g/mol) 289.586
PubChem CID 99728622
Formula Weight 289.59
Percent Purity 96%
Physical Form Viscous Liquid
Chemical Name or Material Methyl 2-chloro-4-(pentafluoroethyl)nicotinate
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.