missing translation for 'onlineSavingsMsg'
Learn More

Methyl 2-chloro-4-fluorophenylacetate, 98%

Product Code. 10492451 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
5 g
Packaging:
Glass Bottle
Unit Size:
5g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10492451 5 g Glass Bottle 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 10492451 Supplier Thermo Scientific Acros Supplier No. 304780050
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 214262-88-1 | C9H8ClFO2 | 202.61 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Specifications

CAS 214262-88-1
Color Yellow
Infrared Spectrum Authentic
Assay Percent Range 98%
Packaging Glass Bottle
Molecular Formula C9H8ClFO2
Linear Formula ClC6H3(F)CHCOCH3
Refractive Index 1.5060 to 1.5080 (20°C, 589nm)
MDL Number MFCD00800611
Quantity 5 g
Synonym methyl 2-2-chloro-4-fluorophenyl acetate, methyl 2-chloro-4-fluorophenylacetate, benzeneacetic acid, 2-chloro-4-fluoro-, methyl ester, methyl-2-chloro-4-fluorophenylacetate, pubchem16488, methyl 2-chloro-4-fluorophenyl acetate, methyl2-2-chloro-4-fluorophenyl acetate, 2-chloro-4-fluorophenyl-acetic acid methyl ester, 2-chloro-4-fluorophenyl acetic acid methyl ester, methyl 2-chloro-4-fluorophenylacetate 98
InChI Key DFTYYBRLWCAUHP-UHFFFAOYSA-N
SMILES COC(=O)CC1=CC=C(F)C=C1Cl
IUPAC Name methyl 2-(2-chloro-4-fluorophenyl)acetate
Molecular Weight (g/mol) 202.61
PubChem CID 736123
Formula Weight 202.51
Percent Purity 98%
Physical Form Liquid
Chemical Name or Material Methyl 2-chloro-4-fluorophenylacetate, 98%
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.