missing translation for 'onlineSavingsMsg'
Learn More

Methyl 2-Bromopropionate, TRC

Product Code. 30469395
Change view
Click to view available options
Quantity:
5 g
Unit Size:
5g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30469395 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30469395 Supplier Toronto Research Chemicals Supplier No. TRCM2940455G
 Restricted Item

This item requires a price update prior to being ordered.

Request a Price for this item here.

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material 2-Bromopropionicacid-methylesterr
CAS 5445-17-0
Molecular Formula C4 H7 Br O2
Quantity 5 g
Synonym Propanoic acid, 2-bromo-, methyl ester, Propionic acid, 2-bromo-, methyl ester (7CI,8CI), (±)-2-Bromopropionic acid methyl ester, (±)-Methyl 2-bromopropionate, 2-Bromopropanoic acid methyl ester, 2-Bromopropionic acid methyl ester, DL-α-Bromopropionic acid methyl ester, Methyl (±)-α-bromopropionate, Methyl 2-bromopropanoate, Methyl 2-bromopropionate, Methyl DL-2-bromopropanoate, Methyl DL-α-bromopropionate, Methyl α-bromopropanoate, Methyl α-bromopropionate, NSC 21973
InChI Formula InChI=1S/C4H7BrO2/c1-3(5)4(6)7-2/h3H,1-2H3
SMILES COC(=O)C(C)Br
IUPAC Name methyl 2-bromopropanoate
Molecular Weight (g/mol) 167
Formula Weight 165.9629
Título del producto
Seleccione un problema

Al hacer clic en Enviar, acepta que Fisher Scientific se ponga en contacto con usted en relación con los comentarios que ha proporcionado en este formulario. No compartiremos su información para ningún otro fin. Toda la información de contacto proporcionada se mantendrá de acuerdo con nuestra Política de Privacidad. Política de privacidad.