missing translation for 'onlineSavingsMsg'
Learn More

Methyl 2-(bromomethyl)acrylate, 96%

Product Code. 11994961 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
Unit Size:
1g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11994961 25 g 25g
10384592 1 g 1g
10456442 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11994961 Supplier Thermo Scientific Acros Supplier No. 367630250
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 4224-69-5 | C5H7BrO2 | 179.01 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 4224-69-5
Molecular Formula C5H7BrO2
Molecular Weight (g/mol) 179.01
MDL Number MFCD00011697
InChI Key CFTUQSLVERGMHL-UHFFFAOYSA-N
Synonym methyl 2-bromomethyl acrylate, methyl 2-bromomethyl prop-2-enoate, methyl 2-bromomethyl-2-propenoate, methyl alpha-bromomethyl acrylate, 2-bromomethyl methyl acrylate, 2-propenoic acid, 2-bromomethyl-, methyl ester, 2-bromomethyl acrylic acid methyl ester, acrylic acid, 2-bromomethyl-, methyl ester, zlchem 195, pubchem17371
PubChem CID 521093
IUPAC Name methyl 2-(bromomethyl)prop-2-enoate
SMILES COC(=O)C(=C)CBr

Specifications

Color Brown to Colorless
Density 1.4890g/mL
Boiling Point 35.0°C to 37.0°C (13.0 mmHg)
Flash Point 78°C
Infrared Spectrum Authentic
Assay Percent Range 95% min. (GC)
Packaging Glass bottle
Refractive Index 1.4870 to 1.4890
Quantity 25 g
Specific Gravity 1.489
Formula Weight 179.01
Percent Purity 96%
Physical Form Liquid
Chemical Name or Material Methyl 2-(bromomethyl)acrylate
Show More Show Less
compliance-icons
Product Identifier
  • Methyl 2-(bromomethyl)acrylate
Signal Word
  • Warning
Hazard Category
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H312-Harmful in contact with skin.
  • H332-Harmful if inhaled.
  • H302-Harmful if swallowed.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.