missing translation for 'onlineSavingsMsg'
Learn More

Methyl 2-bromohexanoate, 98%, Thermo Scientific™

Product Code. 15466638
Change view
Click to view available options
Quantity:
25 mL
Unit Size:
25mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15466638 25 mL 25mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 15466638 Supplier Thermo Scientific Alfa Aesar Supplier No. H60165.AC
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 5445-19-2
Molecular Formula C7H13BrO2
Molecular Weight (g/mol) 209.083
MDL Number MFCD00045038
InChI Key YGLPDRIMFIXNBI-UHFFFAOYSA-N
Synonym 2-bromohexanoic acid methyl ester, hexanoic acid, 2-bromo-, methyl ester, methyl 2-bromocaproate, methyl 2-bromanylhexanoate, methyl bromohexanoate, hexanoic acid, 2-bromo-, methyl ester, s, methyl-2-bromohexanoate, acmc-20mk9n, ksc490a4j, methyl-2-bromocaproate
PubChem CID 95578
IUPAC Name methyl 2-bromohexanoate
SMILES CCCCC(C(=O)OC)Br

Specifications

Density 1.289
Flash Point 69°C (156°F)
Linear Formula CH3(CH2)3CHBrCOOCH3
Refractive Index 1.455
Quantity 25 mL
Beilstein 1722052
Formula Weight 209.08
Percent Purity 98%
Chemical Name or Material Methyl 2-bromohexanoate
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.