missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Methyl 2-bromobenzoate, TRC
Click to view available options
Quantity:
500 mg
Unit Size:
500mg
Specifications
Specifications
| Chemical Name or Material | Methyl 2-bromobenzoate |
| CAS | 610-94-6 |
| Molecular Formula | C8 H7 Br O2 |
| Quantity | 500 mg |
| InChI Formula | InChI=1S/C8H7BrO2/c1-11-8(10)6-4-2-3-5-7(6)9/h2-5H,1H3 |
| SMILES | COC(=O)c1ccccc1Br |
| IUPAC Name | methyl 2-bromobenzoate |
| Molecular Weight (g/mol) | 215.044 |
| Formula Weight | 213.963 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction