missing translation for 'onlineSavingsMsg'
Learn More

Methyl 2-bromobenzoate, 98%

Product Code. 10245761 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
50 mL
Packaging:
Glass bottle
Unit Size:
50mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
10245761 50 mL Glass bottle 50mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 10245761 Supplier Thermo Scientific Acros Supplier No. 249810500
 Restricted Item
Estimated Shipment: 20-10-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 610-94-6 | C8H7BrO2 | 215.05 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Specifications

CAS 610-94-6
Color Yellow
Density 1.5100g/mL
Boiling Point 252.0°C to 254.0°C
Infrared Spectrum Authentic
Assay Percent Range 98%
Packaging Glass bottle
Molecular Formula C8H7BrO2
Linear Formula BrC6H4CO2CH3
Refractive Index 1.5575 to 1.5595
MDL Number MFCD00016328
Quantity 50 mL
Specific Gravity 1.51
Synonym methyl o-bromobenzoate, 2-bromobenzoic acid methyl ester, benzoic acid, 2-bromo-, methyl ester, methyl-2-bromobenzoate, methyl bromobenzoate, o-bromobenzoic acid, methyl ester, benzoic acid, o-bromo-, methyl ester, pubchem3956, 2-bromo methyl benzoate, acmc-1atow
Solubility Information Solubility in water: insoluble. Other solubilities: soluble in polar solvents,insoluble in non-polar solvents
InChI Key SWGQITQOBPXVRC-UHFFFAOYSA-N
SMILES COC(=O)C1=CC=CC=C1Br
IUPAC Name methyl 2-bromobenzoate
Molecular Weight (g/mol) 215.05
PubChem CID 11894
Formula Weight 215.05
Percent Purity 98%
Physical Form Liquid
Chemical Name or Material Methyl 2-bromobenzoate, 98%
Show More Show Less
compliance-icons
Product Identifier
  • Methyl 2-bromobenzoate
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H315-Causes skin irritation.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.