missing translation for 'onlineSavingsMsg'
Learn More

Methyl 2-bromo-4-methylbenzoate, 98%

Product Code. 15419678
Change view
Click to view available options
Quantity:
250 mg
1 g
5 g
Unit Size:
1g
250mg
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15419678 5 g 5g
15429678 250 mg 250mg
15409678 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 15419678 Supplier Thermo Scientific Alfa Aesar Supplier No. H63144.06
 Restricted Item
Estimated Shipment: 04-12-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 87808-49-9 | C9H9BrO2 | 229.073 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 87808-49-9
Molecular Formula C9H9BrO2
Molecular Weight (g/mol) 229.073
MDL Number MFCD03840737
InChI Key DJTUYAFJAGLQCK-UHFFFAOYSA-N
Synonym methyl2-bromo-4-methylbenzoate, 2-bromo-4-methylbenzoic acid methyl ester, benzoic acid, 2-bromo-4-methyl-, methyl ester, pubchem14035, acmc-209qq7, ksc495m0h, methyl 2-bromo-4-methyl-benzoate, 2-bromo-4-methyl-benzoic acid methyl ester
PubChem CID 21779704
IUPAC Name methyl 2-bromo-4-methylbenzoate
SMILES CC1=CC(=C(C=C1)C(=O)OC)Br

Specifications

Quantity 5 g
Formula Weight 229.07
Percent Purity 98%
Physical Form Solid
Chemical Name or Material Methyl 2-bromo-4-methylbenzoate

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.