missing translation for 'onlineSavingsMsg'
Learn More

Methyl 2-bromo-3-methylbenzoate, 98%

Product Code. 15428418
Change view
Click to view available options
Quantity:
25 g
100 g
Unit Size:
100g
25g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15428418 25 g 25g
15438418 100 g 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15428418 Supplier Thermo Scientific Alfa Aesar Supplier No. H61831.14
 Restricted Item
In Stock
Add to basket
This item is not returnable. View return policy

CAS: 131001-86-0 | C9H9BrO2 | 229.07 g/mol

Methyl 2-bromo-3-methylbenzoate is used as pharmaceutical intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Methyl 2-bromo-3-methylbenzoate is used as pharmaceutical intermediate.

Solubility
Slightly soluble in water.

Notes
Stable under recommended storage conditions. Incompatible with oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 131001-86-0
Molecular Formula C9H9BrO2
Molecular Weight (g/mol) 229.07
MDL Number MFCD00144762
InChI Key QAOFGUXVDAZKBW-UHFFFAOYSA-N
Synonym 2-bromo-3-methyl-benzoic acid methyl ester, methyl2-bromo-3-methylbenzoate, benzoic acid, 2-bromo-3-methyl-, methyl ester, 2-bromo-3-methylbenzoic acid methyl ester, methyl 2-bromo-3-methyl-benzoate, pubchem3966, acmc-1c3gk, methyl 2-bromo-m-toluate, intermediates-zcf02010, ksc495c9h
PubChem CID 10823279
IUPAC Name methyl 2-bromo-3-methylbenzoate
SMILES COC(=O)C1=C(Br)C(C)=CC=C1

Specifications

Color Red
Quantity 25 g
Beilstein 4177165
Solubility Information Slightly soluble in water.
Formula Weight 229.07
Percent Purity 98%
Physical Form Liquid
Chemical Name or Material Methyl 2-bromo-3-methylbenzoate
compliance-icons
Product Identifier
  • Methyl 2-bromo-3-methylbenzoate
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/soap.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P312-Call a POISON CENTER/doctor/if you feel unwell.
  • P332+P313-If skin irritation occurs: Get medical advice/attention.
  • P337+P313-If eye irritation persists: Get medical advice/attention.
Supplemental information
  • MIXTURE LIST-Contain: Methyl 2-bromo-3-methylbenzoate

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.