missing translation for 'onlineSavingsMsg'
Learn More

Methyl 2-bromo-3-fluoropropionate, 97%

Product Code. 15444427
Change view
Click to view available options
Quantity:
250 mg
1 g
Unit Size:
1g
250mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15444427 250 mg 250mg
15434427 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15444427 Supplier Thermo Scientific Alfa Aesar Supplier No. H31790.MD
 Restricted Item
Estimated Shipment: 07-12-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 1537-52-6 | C4H6BrFO2 | 184.99 g/mol

In organic synthesis. Employed as important intermediates in various areas such as peptide synthesis, asymmetric synthesis.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
In organic synthesis. Employed as important intermediates in various areas such as peptide synthesis, asymmetric synthesis.

Solubility
Insoluble in water.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from strong oxidizing agents, bases.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1537-52-6
Molecular Formula C4H6BrFO2
Molecular Weight (g/mol) 184.99
MDL Number MFCD09800641
InChI Key BXTYDNLOFHBZFS-UHFFFAOYNA-N
Synonym 2-bromo-3-fluoropropionic acid methyl ester, methyl 2-bromo-3-fluoropropionate, methyl 2-bromo-3-fluoro-propanoate, methyl alpha-bromo-beta-fluoropropionate, methyl 2-bromo-3-fluoroproprionate, propanoic acid, 2-bromo-3-fluoro-, methyl ester
PubChem CID 5271584
IUPAC Name methyl 2-bromo-3-fluoropropanoate
SMILES COC(=O)C(Br)CF

Specifications

Boiling Point 50°C to 54°C (8 mmHg)
Quantity 250 mg
UN Number UN3265
Solubility Information Insoluble in water.
Formula Weight 184.99
Percent Purity 97%
Chemical Name or Material Methyl 2-bromo-3-fluoropropionate

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.