missing translation for 'onlineSavingsMsg'
Learn More

Methyl 2-amino-5-(trifluoromethyl)benzoate, 98%

Product Code. 15431409
Change view
Click to view available options
Quantity:
250 mg
1 g
5 g
Unit Size:
1g
250mg
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15431409 5 g 5g
15488816 250 mg 250mg
15421409 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 15431409 Supplier Thermo Scientific Alfa Aesar Supplier No. H64283.06
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 117324-58-0 | C9H8F3NO2 | 219.163 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 117324-58-0
Molecular Formula C9H8F3NO2
Molecular Weight (g/mol) 219.163
MDL Number MFCD08234902
InChI Key QGFUDNJZHZNPCS-UHFFFAOYSA-N
PubChem CID 14233808
IUPAC Name methyl 2-amino-5-(trifluoromethyl)benzoate
SMILES COC(=O)C1=C(C=CC(=C1)C(F)(F)F)N

Specifications

Color White
Quantity 5 g
Synonym methyl 2-amino-5-trifluoromethyl benzoate, methyl2-amino-5-trifluoromethyl benzoate, 2-amino-5-trifluoromethyl benzoic acid methyl ester, benzoic acid,2-amino-5-trifluoromethyl-, methyl ester, benzoic acid, 2-amino-5-trifluoromethyl-, methyl ester, acmc-209z2m, benzoicacid,2-amino-5-trifluoromethyl-,methylester, methyl 5-trifluoromethylanthranilate, 2-amino-5-trifluoromethyl-benzoic acid methyl ester
Formula Weight 219.16
Percent Purity 98%
Physical Form Solid
Chemical Name or Material Methyl 2-amino-5-(trifluoromethyl)benzoate

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.