missing translation for 'onlineSavingsMsg'
Learn More

Methyl 2-amino-5,6-dihydro-4H-cyclopenta[b]thiophene-3-carboxylate, 96%, Thermo Scientific™

Product Code. 15458455
Change view
Click to view available options
Quantity:
1 g
10 g
Unit Size:
10g
1g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15458455 1 g 1g
15468455 10 g 10g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15458455 Supplier Thermo Scientific Alfa Aesar Supplier No. H33412.03
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 184174-80-9
Molecular Formula C9H11NO2S
Molecular Weight (g/mol) 197.25
MDL Number MFCD00625612
InChI Key XUKGZPUAFGAYHC-UHFFFAOYSA-N
Synonym methyl 2-amino-5,6-dihydro-4h-cyclopenta b thiophene-3-carboxylate, 2-amino-5,6-dihydro-4h-cyclopenta b thiophene-3-carboxylic acid methyl ester, methyl 2-amino-4h,5h,6h-cyclopenta b thiophene-3-carboxylate, methyl 2-amino-5,6-dihydro-4h-cyclopenta b-thiophene-3-carboxylate, 4h-cyclopenta b thiophene-3-carboxylicacid, 2-amino-5,6-dihydro-, methyl ester, methyl 2-amino-4,5,6-trihydrocyclopenta 1,2-b thiophene-3-carboxylate, enamine_005617, 2-amino-5,6-dihydro-4h-cyclopenta b thiophene-3-ca, methyl 2-azanyl-5,6-dihydro-4h-cyclopenta b thiophene-3-carboxylate, 4h-cyclopenta b thiophene-3-carboxylic acid, 2-amino-5,6-dihydro-, methyl
PubChem CID 2818378
IUPAC Name methyl 2-amino-5,6-dihydro-4H-cyclopenta[b]thiophene-3-carboxylate
SMILES COC(=O)C1=C(N)SC2=C1CCC2

Specifications

Melting Point ∽168°C (decomposition)
Quantity 1 g
Formula Weight 197.26
Percent Purity 96%
Chemical Name or Material Methyl 2-amino-5,6-dihydro-4H-cyclopenta[b]thiophene-3-carboxylate
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.