missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Methyl 2-amino-5,6-dihydro-4H-cyclopenta[b]thiophene-3-carboxylate, 95%, Thermo Scientific™
Click to view available options
Quantity:
1 g
10 g
Especificaciones
Melting Point | 170°C to 174°C |
Assay Percent Range | 94% min. (HPLC) |
Quantity | 10 g |
Formula Weight | 197.26 |
Percent Purity | 95% |
Chemical Name or Material | Methyl 2-amino-5, 6-dihydro-4H-cyclopenta[b]thiophene-3-carboxylate |
Seguridad y manipulación
Nombre del producto
- Methyl 2-amino-5,6-dihydro-4H-cyclopenta¢b!thiophene-3-carboxylate, 95%
- Warning
- Serious eye damage/eye irritation Category 2
- Skin corrosion/irritation Category 2
- Specific target organ toxicity Category 3
- H319-Causes serious eye irritation.
- H335-May cause respiratory irritation.
- H315-Causes skin irritation.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P302+P352-IF ON SKIN: Wash with plenty of water/.
- P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P312-Call a POISON CENTER/doctor/if you feel unwell.
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'